Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000029.3 | NP_000020.1 | 3 | 1499 | ||
GenBank mRNA | AB209769.1 | - | 3 | 1011 | ||
AK222798.1 | - | 3 | 1030 | |||
AK222815.1 | - | 3 | 1031 | |||
AK293507.1 | - | 4 | 784 | |||
AK303755.1 | - | 4 | 972 | |||
AK307978.1 | - | 4 | 945 | |||
AK312291.1 | - | 3 | 1109 | |||
BC011519.1 | - | 3 | 1019 | 4213559 | ||
BT006851.1 | - | 2 | 991 | |||
K02215.1 | - | 3 | 1030 | |||
M69110.1 | - | 2 | 1005 |