Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOCS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001075098.3 | NP_001068566.1 | 11 | 2174 | ||
NM_005943.5 | NP_005934.2 | 10 | 2178 | |||
NR_033233.1 | 9 | 1984 | ||||
XM_011514632.2 | XP_011512934.1 | 11 | 2070 | |||
XM_011514633.2 | XP_011512935.1 | 10 | 2022 | |||
XM_011514634.2 | XP_011512936.1 | 10 | 1967 | |||
XM_017010888.1 | XP_016866377.1 | 10 | 1992 | |||
GenBank mRNA | AF034374.1 | - | 11 | 2057 | ||
AJ224328.1 | - | 10 | 2175 | |||
AK056740.1 | - | 9 | 1930 | |||
AK091306.1 | - | 10 | 1906 |