Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCIRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006019.3 | NP_006010.2 | 16, 17 | 2142 | ||
NM_006053.3 | NP_006044.1 | 11, 12 | 1901 | |||
XM_005273709.3 | XP_005273766.1 | 17, 18 | 2301 | |||
XM_011544726.2 | XP_011543028.1 | 16, 17 | 2041 | |||
XM_017017089.1 | XP_016872578.1 | 16, 17 | 2137 | |||
XM_017017090.1 | XP_016872579.1 | 15, 16 | 2480 | |||
GenBank mRNA | AF025374.1 | - | 11, 12 | 1901 | ||
AY548970.1 | - | 1, 2 | 641 | |||
BC018133.1 | - | 16, 17 | 2121 | 3905446 | ||
BC032465.1 | - | 16, 17 | 2114 | 5210733 | ||
U45285.1 | - | 16, 17 | 2067 |