Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGCX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000821.6 | NP_000812.2 | 4 | 588 | ||
NM_001142269.3 | NP_001135741.1 | 3 | 417 | |||
XM_005264259.4 | XP_005264316.1 | 4 | 547 | |||
XM_011532764.2 | XP_011531066.1 | 4 | 547 | |||
XM_011532765.2 | XP_011531067.1 | 3 | 386 | |||
XM_017003803.1 | XP_016859292.1 | 3 | 410 | |||
GenBank mRNA | AK222827.1 | - | 4 | 548 | ||
AK225663.1 | - | 4 | 547 | |||
AK297397.1 | - | 3 | 357 | |||
AK313126.1 | - | 4 | 556 | |||
BC013979.2 | - | 4 | 510 | 4129593 | ||
EU847509.1 | - | 4 | 485 | |||
L17128.1 | - | 4 | 637 | |||
M81592.1 | - | 4 | 568 |