Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYBB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000397.3 | NP_000388.2 | 13 | 1699 | ||
GenBank mRNA | AK289753.1 | - | 13 | 1684 | ||
AK301476.1 | - | 11 | 1306 | |||
AK301491.1 | - | 12 | 1561 | |||
AK312929.1 | - | 13 | 1660 | |||
BC032720.1 | - | 13 | 1685 | 5505170 | ||
KU178009.1 | - | 13 | 1638 | |||
KU178010.1 | - | 7 | 611 | |||
X04011.1 | - | 13 | 1653 |