Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP3A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202855.2 | NP_001189784.1 | 2, 3 | 281 | ||
NM_017460.5 | NP_059488.2 | 2, 3 | 281 | |||
XM_017011780.1 | XP_016867269.1 | 2, 3 | 279 | |||
GenBank mRNA | AF182273.1 | - | 2, 3 | 183 | ||
AK298451.1 | - | 2, 3 | 274 | |||
AK298462.1 | - | 2, 3 | 280 | |||
AK312967.1 | - | 2, 3 | 274 | |||
AY606313.2 | - | 2, 3 | 174 | |||
BC069418.1 | - | 2, 3 | 224 | 7262313 | ||
BC101631.1 | - | 2, 3 | 270 | 8069137 | ||
D00003.1 | - | 2, 3 | 234 | |||
DA639071.1 | - | 2, 3 | 359 | |||
DQ924960.1 | - | 2, 3 | 174 | |||
J04449.1 | - | 2, 3 | 242 | |||
M13785.1 | - | 2, 3 | 234 | |||
M14096.1 | - | 2, 3 | 242 | |||
M18907.1 | - | 2, 3 | 264 | |||
X12387.1 | - | 2, 3 | 243 |