Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSEN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145392.1 | NP_001138864.1 | 5 | 533 | ||
NM_001145393.2 | NP_001138865.1 | 5 | 501 | |||
NM_001145394.1 | NP_001138866.1 | 5 | 501 | |||
NM_001321277.1 | NP_001308206.1 | 5 | 501 | |||
NM_001321278.1 | NP_001308207.1 | 5 | 501 | |||
NM_001321279.1 | NP_001308208.1 | 5 | 767 | |||
NM_025265.3 | NP_079541.1 | 5 | 785 | |||
NR_135607.1 | 5 | 501 | ||||
XM_011534139.2 | XP_011532441.1 | 3 | 684 | |||
XM_017007292.1 | XP_016862781.1 | 6 | 510 | |||
XM_017007293.1 | XP_016862782.1 | 6 | 842 | |||
XM_017007294.1 | XP_016862783.1 | 6 | 576 | |||
XM_017007295.1 | XP_016862784.1 | 6 | 772 | |||
XM_017007296.1 | XP_016862785.1 | 5 | 1081 | |||
XM_017007297.1 | XP_016862786.1 | 6 | 842 | |||
GenBank mRNA | AK027821.1 | - | 5 | 784 | ||
AK074794.1 | - | 5 | 432 | |||
AK225875.1 | - | 5 | 490 | |||
AK291183.1 | - | 5 | 570 | |||
AK295508.1 | - | 4 | 660 | |||
AK300449.1 | - | 5 | 750 | |||
BC004178.2 | - | 5 | 518 | 2959536 | ||
BC004211.2 | - | 5 | 518 | 2959536 | ||
BC019582.1 | - | 5 | 767 | 3872331 | ||
CN341716.1 | - | 4 | 238 | |||
DA614108.1 | - | 5 | 502 |