Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4orf26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206981.1 | NP_001193910.1 | 3 | 911 | ||
NM_001257072.1 | NP_001244001.1 | 3 | 732 | |||
NM_178497.3 | NP_848592.2 | 2 | 867 | |||
NR_046429.1 | 5 | 887 | ||||
NR_046430.1 | 4 | 868 | ||||
GenBank mRNA | AK300227.1 | - | 3 | 894 |