Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4orf26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206981.1 | NP_001193910.1 | 3 | 671 | ||
NM_001257072.1 | NP_001244001.1 | 3 | 492 | |||
NM_178497.3 | NP_848592.2 | 2 | 627 | |||
NR_046429.1 | 5 | 647 | ||||
NR_046430.1 | 4 | 628 | ||||
XM_011531668.2 | XP_011529970.1 | 4 | 1626 | |||
GenBank mRNA | AK074237.1 | - | 2 | 627 | ||
AK172776.1 | - | 2 | 627 | |||
AK300227.1 | - | 3 | 654 |