Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OXCT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000436.3 | NP_000427.1 | 5 | 849 | ||
XM_017009494.1 | XP_016864983.1 | 5 | 730 | |||
GenBank mRNA | AK312327.1 | - | 5 | 545 | ||
BC009001.1 | - | 5 | 573 | 4148643 | ||
CR457049.1 | - | 5 | 518 | |||
EF095213.1 | - | 2 | 255 | |||
U62961.1 | - | 5 | 616 |