Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC34A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177998.1 | NP_001171469.1 | 8 | 950 | ||
NM_001177999.1 | NP_001171470.1 | 8 | 973 | |||
NM_006424.2 | NP_006415.2 | 8 | 953 | |||
GenBank mRNA | AF111856.1 | - | 8 | 935 | ||
AF146796.1 | - | 8 | 904 | |||
AK075015.1 | - | 2 | 79 | |||
AK075046.1 | - | 8 | 915 | |||
AK292675.1 | - | 8 | 941 | |||
AK314514.1 | - | 8 | 962 | |||
BC142704.1 | - | 8 | 898 | 40148805 | ||
BC146666.1 | - | 8 | 898 | 40148124 | ||
KM822236.1 | - | 7 | 872 |