Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC34A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177998.1 | NP_001171469.1 | 10 | 1222 | ||
NM_001177999.1 | NP_001171470.1 | 10 | 1245 | |||
NM_006424.2 | NP_006415.2 | 10 | 1225 | |||
GenBank mRNA | AF111856.1 | - | 10 | 1207 | ||
AF146796.1 | - | 10 | 1176 | |||
AK075015.1 | - | 4 | 351 | |||
AK075046.1 | - | 10 | 1187 | |||
AK292675.1 | - | 10 | 1213 | |||
AK314514.1 | - | 10 | 1234 | |||
BC142704.1 | - | 10 | 1170 | 40148805 | ||
BC146666.1 | - | 10 | 1170 | 40148124 | ||
KM822236.1 | - | 9 | 1144 |