Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007374.2 | NP_031400.2 | 1 | 701 | ||
GenBank mRNA | AJ011785.1 | - | 1 | 596 | ||
BC065831.1 | - | 1 | 701 | |||
BC069413.1 | - | 1 | 605 | 7262276 | ||
BC087852.1 | - | 1 | 679 |