Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006517.4 | NP_006508.2 | 6 | 1642 | ||
GenBank mRNA | AB085789.1 | - | 6 | 1584 | ||
AK308542.1 | - | 3 | 332 | |||
U05315.1 | - | 6 | 1674 |