Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASQ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001232.3 | NP_001223.2 | 3 | 607 | ||
GenBank mRNA | AK129891.1 | - | 3 | 582 | ||
AK225959.1 | - | 3 | 469 | |||
AK293595.1 | - | 3 | 616 | |||
AK313041.1 | - | 3 | 553 | |||
BC022288.1 | - | 3 | 516 | 4289141 | ||
D55655.1 | - | 3 | 452 |