Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASAH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127505.2 | NP_001120977.1 | 12 | 1120 | ||
NM_004315.5 | NP_004306.3 | 12 | 1186 | |||
NM_177924.4 | NP_808592.2 | 12 | 1253 | |||
XM_005273504.3 | XP_005273561.1 | 12 | 1014 | |||
GenBank mRNA | AK025732.1 | - | 13 | 1156 | ||
AK098299.1 | - | 12 | 901 | |||
AK222780.1 | - | 13 | 989 | |||
AK289481.1 | - | 13 | 1154 | |||
AY305384.1 | - | 12 | 1027 | |||
BC016481.1 | - | 13 | 976 | 3923451 | ||
BC016828.1 | - | 12 | 1107 | 4109317 | ||
BC035453.1 | - | 7 | 551 | |||
U47674.1 | - | 12 | 957 | |||
U70063.1 | - | 13 | 958 |