Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005270.4 | NP_005261.2 | 11 | 1855 | ||
XM_006712422.2 | XP_006712485.1 | 11 | 1804 | |||
XM_011510969.2 | XP_011509271.2 | 11 | 2487 | |||
XM_011510970.2 | XP_011509272.1 | 11 | 3498 | |||
XM_011510971.2 | XP_011509273.1 | 11 | 2748 | |||
XM_011510972.2 | XP_011509274.2 | 11 | 1885 | |||
XM_011510973.2 | XP_011509275.1 | 10 | 1636 | |||
XM_011510974.2 | XP_011509276.1 | 10 | 1644 | |||
XM_017003818.1 | XP_016859307.1 | 11 | 2436 | |||
GenBank mRNA | AB007295.1 | - | 8 | 1412 | ||
AB007296.1 | - | 8 | 1361 | |||
AB007297.1 | - | 8 | 1412 | |||
AB007298.1 | - | 8 | 1361 | |||
AB209354.1 | - | 3 | 432 | |||
BC111410.1 | - | 11 | 1799 | |||
D14827.1 | - | 8 | 1361 | |||
D14828.1 | - | 7 | 1257 | |||
DQ086814.1 | - | 11 | 1825 |