Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DFNB59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042702.3 | NP_001036167.1 | 7 | 1243 | ||
XM_005246627.2 | XP_005246684.1 | 7 | 1468 | |||
XM_005246628.3 | XP_005246685.1 | 6 | 1515 | |||
XM_005246629.3 | XP_005246686.1 | 7 | 1367 | |||
XM_011511247.2 | XP_011509549.1 | 7 | 1588 | |||
XM_011511249.2 | XP_011509551.1 | 6 | 876 | |||
XM_011511250.2 | XP_011509552.1 | 7 | 1245 | |||
XM_011511251.1 | XP_011509553.1 | 6 | 693 | |||
XM_017004221.1 | XP_016859710.1 | 7 | 1614 | |||
XM_017004222.1 | XP_016859711.1 | 7 | 1782 | |||
XM_017004223.1 | XP_016859712.1 | 6 | 1817 | |||
XM_017004224.1 | XP_016859713.1 | 6 | 2371 | |||
XM_017004225.1 | XP_016859714.1 | 6 | 953 | |||
GenBank mRNA | BC127902.1 | - | 6 | 919 | 40132882 | |
BC127903.1 | - | 5 | 1630 | 40132890 | ||
BC146938.1 | - | 7 | 1019 | 9056866 | ||
DQ365827.1 | - | 7 | 1243 |