Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256267.1 | NP_001243196.1 | 13 | 2770 | ||
NM_001256268.1 | NP_001243197.1 | 16 | 3179 | |||
NM_032578.3 | NP_115967.2 | 12 | 2875 | |||
NR_045662.3 | 12 | 2010 | ||||
XM_017016833.1 | XP_016872322.1 | 13 | 3211 | |||
XM_017016834.1 | XP_016872323.1 | 12 | 2669 | |||
GenBank mRNA | AF328296.1 | - | 12 | 3070 | ||
AL832002.2 | - | 12 | 2798 | |||
AL832379.1 | - | 12 | 1972 | |||
AL834247.2 | - | 12 | 2797 | |||
BX647637.1 | - | 12 | 2807 | |||
BX647640.1 | - | 13 | 2763 |