Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005105.4 | NP_005096.1 | 2 | 190 | ||
GenBank mRNA | AF127761.1 | - | 2 | 92 | ||
AF161463.1 | - | 2 | 104 | |||
AF182415.1 | - | 2 | 109 | |||
AF198620.1 | - | 2 | 114 | |||
AF231511.1 | - | 2 | 100 | |||
AF299118.1 | - | 2 | 80 | |||
AK075009.1 | - | 2 | 111 | |||
BC017088.2 | - | 2 | 152 | 3850355 | ||
BC017770.1 | - | 2 | 109 | |||
BC071577.1 | - | 2 | 138 | |||
CR541805.1 | - | 2 | 80 | |||
CR541823.1 | - | 2 | 80 | |||
DB286556.1 | - | 2 | 190 | |||
KF051020.1 | - | 2 | 80 |