Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020919.3 | NP_065970.2 | 13, 14 | 2945 | ||
XM_006712654.2 | XP_006712717.1 | 13, 14 | 3522 | |||
XM_006712655.3 | XP_006712718.1 | 9, 10 | 1159 | |||
XM_011511530.2 | XP_011509832.1 | 12, 13 | 2657 | |||
XM_017004569.1 | XP_016860058.1 | 12, 13 | 2649 | |||
XM_017004570.1 | XP_016860059.1 | 12, 13 | 2649 | |||
XM_017004571.1 | XP_016860060.1 | 12, 13 | 2587 | |||
XM_017004572.1 | XP_016860061.1 | 10, 11 | 2237 | |||
GenBank mRNA | AB046783.2 | - | 12, 13 | 2666 | ||
AB053305.1 | - | 13, 14 | 2712 | |||
AF391100.1 | - | 13, 14 | 2712 | |||
AK056413.1 | - | 4, 5 | 550 | |||
AK291029.1 | - | 13, 14 | 2729 | |||
BC071576.1 | - | 13, 14 | 2707 | 30347073 |