Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020944.2 | NP_065995.1 | 2 | 946 | ||
XM_005251526.4 | XP_005251583.1 | 2 | 973 | |||
XM_006716809.3 | XP_006716872.1 | 2 | 973 | |||
XM_011517973.2 | XP_011516275.1 | 2 | 551 | |||
XM_017014937.1 | XP_016870426.1 | 2 | 973 | |||
XM_017014938.1 | XP_016870427.1 | 2 | 973 | |||
XM_017014939.1 | XP_016870428.1 | 2 | 551 | |||
XM_017014940.1 | XP_016870429.1 | 2 | 551 | |||
XM_017014941.1 | XP_016870430.1 | 2 | 973 | |||
XM_017014942.1 | XP_016870431.1 | 2 | 551 | |||
XM_017014943.1 | XP_016870432.1 | 2 | 551 | |||
XM_017014946.1 | XP_016870435.1 | 2 | 551 | |||
GenBank mRNA | AB046825.1 | - | 2 | 936 | ||
AJ309567.1 | - | 2 | 946 | |||
AK027884.1 | - | 2 | 609 | |||
AK295967.1 | - | 3 | 921 | |||
AL834306.1 | - | 2 | 706 | |||
BC011363.1 | - | 2 | 905 | 4339156 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AF334829.3 | - | 1 | 46814 | Chr. 9 - 35605184 - 35791373 [-] on Build GRCh38 |