Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARID1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006015.4 | NP_006006.3 | 20 | 6460 | ||
NM_139135.2 | NP_624361.1 | 20 | 5809 | |||
GenBank mRNA | AF219114.1 | - | 20 | 5049 | ||
AF231056.1 | - | 20 | 6465 | |||
AF265208.1 | - | 20 | 5015 | |||
AF268913.1 | - | 19 | 4287 | |||
AF521670.1 | - | 20 | 5229 | |||
AK027442.1 | - | 1 | 964 | |||
AK027467.1 | - | 1 | 798 | |||
AK027655.1 | - | 6 | 1504 | |||
AK074940.1 | - | 3 | 1519 | |||
AK223275.1 | - | 9 | 2777 | |||
AK308363.1 | - | 4 | 1585 |