Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL1RN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318914.1 | NP_001305843.1 | 1, 2 | 137 | ||
NM_173841.2 | NP_776213.1 | 1, 2 | 137 | |||
GenBank mRNA | AK290898.1 | - | 1, 2 | 127 | ||
DA667440.1 | - | 1, 2 | 54 | |||
X84348.1 | - | 1, 2 | 43 |