Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152743.3 | NP_689956.2 | 2 | 401 | ||
XM_005249643.4 | XP_005249700.1 | 2 | 363 | |||
XM_011515177.2 | XP_011513479.1 | 2 | 368 | |||
XM_011515178.1 | XP_011513480.1 | 2 | 579 | |||
XM_011515179.2 | XP_011513481.1 | 2 | 362 | |||
XM_011515181.2 | XP_011513483.1 | 2 | 359 | |||
XM_011515182.2 | XP_011513484.1 | 2 | 154 | |||
XM_011515183.1 | XP_011513485.1 | 2 | 396 | |||
XM_011515184.2 | XP_011513486.1 | 1 | 95 | |||
XM_011515186.2 | XP_011513488.1 | 2 | 360 | |||
XM_017011833.1 | XP_016867322.1 | 2 | 167 | |||
XM_017011834.1 | XP_016867323.1 | 2 | 358 | |||
XM_017011835.1 | XP_016867324.1 | 2 | 396 | |||
XM_017011836.1 | XP_016867325.1 | 2 | 357 | |||
GenBank mRNA | AK024482.1 | - | 2 | 149 | ||
AK131083.1 | - | 2 | 337 | |||
BC015632.2 | - | 2 | 155 | 3839985 | ||
BP363179.1 | - | 2 | 400 |