Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007214.4 | NP_009145.1 | 19 | 2292 | ||
XM_011535399.2 | XP_011533701.1 | 19 | 2041 | |||
XM_017010218.1 | XP_016865707.1 | 19 | 2121 | |||
GenBank mRNA | AF100141.1 | - | 19 | 2023 | ||
AJ011779.1 | - | 19 | 2121 | |||
AJ420532.1 | - | 1 | 833 | |||
AK074835.1 | - | 19 | 2108 | |||
AL137338.1 | - | 16 | 1639 | |||
BC047221.1 | - | 19 | 2161 | 5277083 | ||
BC048287.1 | - | 19 | 2148 | 5212007 | ||
KU178551.1 | - | 19 | 2023 | |||
KU178552.1 | - | 8 | 712 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK125856.1 | - | 1841 | Chr. - - [-] on Build |