Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007214.4 | NP_009145.1 | 18 | 2126 | ||
XM_011535399.2 | XP_011533701.1 | 18 | 1875 | |||
XM_017010218.1 | XP_016865707.1 | 18 | 1955 | |||
GenBank mRNA | AF100141.1 | - | 18 | 1857 | ||
AJ011779.1 | - | 18 | 1955 | |||
AK027374.1 | - | 15 | 1498 | |||
AK074835.1 | - | 18 | 1942 | |||
AL137338.1 | - | 15 | 1473 | |||
BC047221.1 | - | 18 | 1995 | 5277083 | ||
BC048287.1 | - | 18 | 1982 | 5212007 | ||
KU178551.1 | - | 18 | 1857 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK125856.1 | - | 1675 | Chr. - - [-] on Build |