Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000229.1 | NP_000220.1 | 6 | 953 | ||
GenBank mRNA | BC014781.1 | - | 6 | 943 | 4155094 | |
BT009748.1 | - | 6 | 942 | |||
M12625.1 | - | 6 | 1209 | |||
M26268.1 | - | 6 | 895 | |||
X06537.1 | - | 6 | 1045 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE840182.1 | - | 1 | 462 | Chr. 16 - 67940159 - 67940712 [-] on Build GRCh38 |