Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPRSS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289000.1 | NP_001275929.1 | 3 | 312 | ||
NM_001289001.1 | NP_001275930.1 | 3 | 322 | |||
NM_153609.3 | NP_705837.1 | 3 | 391 | |||
XM_011529989.2 | XP_011528291.1 | 3 | 450 | |||
GenBank mRNA | AJ319876.1 | - | 2 | 254 | ||
AY055384.1 | - | 3 | 391 | |||
AY358398.1 | - | 3 | 322 | |||
BC039082.1 | - | 3 | 394 | 5164881 | ||
CR456446.1 | - | 3 | 312 |