Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C12orf76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207435.2 | NP_997318.1 | 3 | 425 | ||
XM_017019297.1 | XP_016874786.1 | 2 | 93 | |||
GenBank mRNA | AK097461.1 | - | 3 | 425 | ||
BC133010.1 | - | 2 | 208 | 40147032 | ||
BC133012.1 | - | 2 | 208 | 40147034 |