Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003793.3 | NP_003784.2 | 5 | 739 | ||
XM_011545328.2 | XP_011543630.1 | 5 | 489 | |||
GenBank mRNA | AF071748.1 | - | 3 | 303 | ||
AF071749.1 | - | 3 | 205 | |||
AF088886.2 | - | 5 | 739 | |||
AF136279.1 | - | 5 | 738 | |||
AJ007331.1 | - | 5 | 694 | |||
AK313657.1 | - | 5 | 707 | |||
AL137742.1 | - | 3 | 170 | |||
BC011682.2 | - | 5 | 733 | 3535532 | ||
BC036451.1 | - | 5 | 714 | 4830274 | ||
CR541680.1 | - | 3 | 211 |