Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYBB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000496.2 | NP_000487.1 | 2, 3 | 112 | ||
XM_006724141.3 | XP_006724204.1 | 2, 3 | 1271 | |||
XM_011529900.2 | XP_011528202.1 | 3, 4 | 1636 | |||
GenBank mRNA | BC069535.1 | - | 1, 2 | 76 | 7262188 | |
CR456426.1 | - | 1, 2 | 74 | |||
KR052782.1 | - | 2, 3 | 106 | |||
KR052783.1 | - | 2, 3 | 106 | |||
KR052784.1 | - | 2, 3 | 108 | |||
KR052788.1 | - | 1, 2 | 64 | |||
KR052789.1 | - | 1, 2 | 55 | |||
KR052790.1 | - | 1, 2 | 97 | |||
KR052791.1 | - | 12 | ||||
L10035.1 | - | 1, 2 | 76 |