Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040875.2 | NP_001035808.1 | 6 | 798 | ||
NM_018972.2 | NP_061845.2 | 6 | 928 | |||
NR_046346.1 | 6 | 783 | ||||
XM_011517551.2 | XP_011515853.1 | 5 | 1886 | |||
XM_011517552.2 | XP_011515854.1 | 5 | 723 | |||
GenBank mRNA | AB551556.1 | - | 6 | 927 | ||
AB551557.1 | - | 6 | 927 | |||
AB551558.1 | - | 6 | 925 | |||
AK292572.1 | - | 6 | 867 | |||
AK295594.1 | - | 6 | 722 | |||
BC024939.1 | - | 6 | 763 | 3923335 | ||
BC035450.1 | - | 6 | 775 | |||
Y17849.1 | - | 6 | 856 |