Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040875.2 | NP_001035808.1 | 6 | 2672 | ||
NM_018972.2 | NP_061845.2 | 6 | 2802 | |||
NR_046346.1 | 6 | 2657 | ||||
XM_011517551.2 | XP_011515853.1 | 5 | 3760 | |||
XM_011517552.2 | XP_011515854.1 | 5 | 2597 | |||
GenBank mRNA | AB551556.1 | - | 6 | 2801 | ||
AB551557.1 | - | 6 | 2801 | |||
AB551558.1 | - | 6 | 2799 | |||
AL110252.1 | - | 1 | 1302 |