Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL17RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289905.1 | NP_001276834.1 | 12 | 1529 | ||
NM_014339.6 | NP_055154.3 | 13 | 1631 | |||
GenBank mRNA | BC011624.2 | - | 13 | 1575 | 4053746 | |
CT841520.1 | - | 13 | 1554 | |||
JF305973.1 | - | 13 | 1498 | |||
U58917.1 | - | 13 | 1530 |