Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNGB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135639.1 | NP_001129111.1 | 6, 7 | 486 | ||
NM_001286130.1 | NP_001273059.1 | 6, 7 | 486 | |||
NM_001297.4 | NP_001288.3 | 6, 7 | 486 | |||
GenBank mRNA | AF042498.1 | - | 6, 7 | 491 | ||
BQ638084.1 | - | 6, 7 | 482 | |||
U18945.1 | - | 6, 7 | 489 | |||
U58837.1 | - | 6, 7 | 478 |