Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199835.1 | NP_001186764.1 | 6 | 684 | ||
NM_001199837.1 | NP_001186766.1 | 4 | 489 | |||
NM_001199838.1 | NP_001186767.1 | 3 | 388 | |||
NM_001318198.1 | NP_001305127.1 | 7 | 758 | |||
NM_001318199.1 | NP_001305128.1 | 6 | 559 | |||
NM_013322.2 | NP_037454.2 | 6 | 637 | |||
XM_006715712.2 | XP_006715775.1 | 6 | 837 | |||
XM_017012085.1 | XP_016867574.1 | 8 | 852 | |||
XM_017012086.1 | XP_016867575.1 | 8 | 1291 | |||
XM_017012087.1 | XP_016867576.1 | 7 | 781 | |||
XM_017012088.1 | XP_016867577.1 | 7 | 728 | |||
GenBank mRNA | AF121860.1 | - | 6 | 550 | ||
AK296142.1 | - | 7 | 588 | |||
AK307905.1 | - | 4 | 438 | |||
AK309162.1 | - | 3 | 388 | |||
AK311256.1 | - | 2 | 448 | |||
AK312850.1 | - | 6 | 559 | |||
AK316088.1 | - | 7 | 734 | |||
BC031050.1 | - | 6 | 531 | 5278015 | ||
BC034992.1 | - | 6 | 610 | 4827599 | ||
BG399132.1 | - | 3 | 401 | |||
BX648363.1 | - | 1 | 1640 | |||
DA377368.1 | - | 6 | 559 |