Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199835.1 | NP_001186764.1 | 6 | 705 | ||
NM_001199837.1 | NP_001186766.1 | 4 | 510 | |||
NM_001199838.1 | NP_001186767.1 | 3 | 409 | |||
NM_001318198.1 | NP_001305127.1 | 7 | 779 | |||
NM_001318199.1 | NP_001305128.1 | 6 | 580 | |||
NM_013322.2 | NP_037454.2 | 6 | 658 | |||
XM_006715712.2 | XP_006715775.1 | 6 | 858 | |||
XM_017012085.1 | XP_016867574.1 | 8 | 873 | |||
XM_017012086.1 | XP_016867575.1 | 8 | 1312 | |||
XM_017012087.1 | XP_016867576.1 | 7 | 802 | |||
XM_017012088.1 | XP_016867577.1 | 7 | 749 | |||
GenBank mRNA | AF121860.1 | - | 6 | 571 | ||
AK296142.1 | - | 7 | 609 | |||
AK307905.1 | - | 4 | 459 | |||
AK309162.1 | - | 3 | 409 | |||
AK311256.1 | - | 2 | 469 | |||
AK312850.1 | - | 6 | 580 | |||
AK316088.1 | - | 7 | 755 | |||
BC031050.1 | - | 6 | 552 | 5278015 | ||
BC034992.1 | - | 6 | 631 | 4827599 | ||
BG399132.1 | - | 3 | 422 | |||
BX648363.1 | - | 1 | 1661 |