Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE6H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006205.2 | NP_006196.1 | 3 | 253 | ||
XM_017019431.1 | XP_016874920.1 | 2 | 496 | |||
GenBank mRNA | BC069774.1 | - | 2 | 190 | 7262475 | |
BC069800.1 | - | 2 | 190 | 7262487 | ||
BC093738.1 | - | 2 | 180 | 7939583 | ||
BC093740.1 | - | 2 | 180 | 7939585 | ||
D45399.1 | - | 3 | 231 | |||
EL946662.1 | - | 3 | 257 |