Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173689.6 | NP_775960.4 | 13 | 4877 | ||
XM_005251934.2 | XP_005251991.1 | 8 | 3897 | |||
XM_011518556.2 | XP_011516858.1 | 13 | 4850 | |||
XM_011518557.2 | XP_011516859.1 | 13 | 4879 | |||
XM_011518558.2 | XP_011516860.1 | 14 | 5239 | |||
GenBank mRNA | AK095783.1 | - | 4 | 1514 | ||
AK124819.1 | - | 1 | 948 |