Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VEGFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025366.2 | NP_001020537.2 | 7 | 1694 | ||
NM_001025367.2 | NP_001020538.2 | 7 | 1625 | |||
NM_001025368.2 | NP_001020539.2 | 6 | 1571 | |||
NM_001033756.2 | NP_001028928.1 | 6 | 1571 | |||
NM_001171623.1 | NP_001165094.1 | 7 | 1694 | |||
NM_001171624.1 | NP_001165095.1 | 7 | 1643 | |||
NM_001171625.1 | NP_001165096.1 | 7 | 1625 | |||
NM_001171626.1 | NP_001165097.1 | 6 | 1571 | |||
NM_001171629.1 | NP_001165100.1 | 6 | 1571 | |||
NM_001287044.1 | NP_001273973.1 | 6 | 584 | |||
NM_001317010.1 | NP_001303939.1 | 6 | 1571 | |||
NM_003376.5 | NP_003367.4 | 7 | 1643 | |||
GenBank mRNA | AB021221.1 | - | 6 | 533 | ||
AB209485.1 | - | 3 | 7752 | |||
AB451322.1 | - | 6 | 533 | |||
AB451451.1 | - | 6 | 533 | |||
AF022375.1 | - | 6 | 1234 | |||
AF062645.1 | - | 4 | 248 | |||
AF430806.1 | - | 6 | 533 | |||
AF486837.1 | - | 6 | 533 | |||
AJ010438.1 | - | 7 | 587 | |||
AY047581.1 | - | 6 | 549 | |||
AY500353.1 | - | 6 | 532 | |||
AY766116.1 | - | 6 | 533 | |||
BQ880667.1 | - | 1 | 111 | |||
BX442364.2 | - | 6 | 576 | |||
KT581011.1 | - | 6 | 533 | |||
M27281.1 | - | 7 | 761 | |||
M32977.1 | - | 6 | 589 | |||
S85192.1 | - | 7 | 724 | |||
X62568.1 | - | 6 | 549 |