Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOTCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200001.1 | NP_001186930.1 | 18 | 3237 | ||
NM_024408.3 | NP_077719.2 | 18 | 3237 | |||
XM_005270901.3 | XP_005270958.1 | 19 | 5551 | |||
XM_011541519.2 | XP_011539821.1 | 18 | 8345 | |||
XM_011541520.2 | XP_011539822.1 | 18 | 5255 | |||
XM_017001372.1 | XP_016856861.1 | 18 | 2907 | |||
XM_017001373.1 | XP_016856862.1 | 18 | 2886 | |||
GenBank mRNA | AF308601.1 | - | 18 | 2952 | ||
AF315356.1 | - | 18 | 2952 | |||
BC071562.1 | - | 18 | 3186 | 4822955 | ||
U77493.1 | - | 1 | 42 |