Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOTCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200001.1 | NP_001186930.1 | 20 | 3531 | ||
NM_024408.3 | NP_077719.2 | 20 | 3531 | |||
XM_005270901.3 | XP_005270958.1 | 21 | 5845 | |||
XM_011541519.2 | XP_011539821.1 | 20 | 8639 | |||
XM_011541520.2 | XP_011539822.1 | 20 | 5549 | |||
XM_017001372.1 | XP_016856861.1 | 20 | 3201 | |||
XM_017001373.1 | XP_016856862.1 | 20 | 3180 | |||
GenBank mRNA | AF315356.1 | - | 20 | 3246 | ||
BC071562.1 | - | 20 | 3480 | 4822955 | ||
U77493.1 | - | 3 | 336 |