Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018060.3 | NP_060530.3 | 23 | 3020 | ||
GenBank mRNA | AK001188.1 | - | 21 | 2494 | ||
AK001726.1 | - | 17 | 2008 | |||
AK022665.1 | - | 13 | 1587 | |||
AK291432.1 | - | 23 | 2829 | |||
AY267462.1 | - | 23 | 3031 | |||
BC010218.1 | - | 19 | 2200 | |||
BC040376.1 | - | 23 | 2753 | |||
BC047880.1 | - | 22 | 2547 | |||
BC137438.1 | - | 23 | 2990 | 9021436 | ||
D28500.1 | - | 23 | 2861 |