Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145038.4 | NP_659475.2 | 5 | 659 | ||
XM_005264637.3 | XP_005264694.1 | 4 | 467 | |||
XM_005264638.3 | XP_005264695.1 | 2 | 154 | |||
XM_017005271.1 | XP_016860760.1 | 5 | 567 | |||
GenBank mRNA | AK057222.1 | - | 5 | 655 | ||
AK289953.1 | - | 5 | 642 | |||
AK307094.1 | - | 4 | 449 | |||
AL833892.1 | - | 4 | 393 | |||
BC009488.2 | - | 4 | 382 |