Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005264638.3 | XP_005264695.1 | 4, 5 | 469 | ||
XM_017005271.1 | XP_016860760.1 | 7, 8 | 882 | |||
GenBank mRNA | AK307094.1 | - | 6, 7 | 764 |