Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLOC1S6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001311255.1 | NP_001298184.1 | 2 | 326 | ||
NM_001311256.1 | NP_001298185.1 | 2 | 326 | |||
NM_012388.3 | NP_036520.1 | 2 | 492 | |||
NR_132350.1 | 2 | 326 | ||||
NR_132351.1 | 2 | 492 | ||||
NR_132352.1 | 2 | 492 | ||||
NR_132356.1 | 2 | 194 | ||||
NR_132357.1 | 2 | 194 | ||||
GenBank mRNA | AF080470.1 | - | 2 | 396 | ||
AK057545.1 | - | 2 | 236 | |||
AK128626.1 | - | 2 | 295 | |||
AL596952.1 | - | 2 | 265 | |||
BC004819.1 | - | 2 | 213 | 3450397 | ||
DA247079.1 | - | 2 | 212 | |||
DA468279.1 | - | 2 | 492 | |||
DA523523.1 | - | 2 | 140 | |||
DA859617.1 | - | 2 | 142 | |||
DB289156.1 | - | 2 | 137 |