Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLOC1S6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001311255.1 | NP_001298184.1 | 3 | 450 | ||
NM_012388.3 | NP_036520.1 | 3 | 616 | |||
NR_132350.1 | 4 | 624 | ||||
NR_132351.1 | 3 | 612 | ||||
NR_132353.1 | 2 | 474 | ||||
NR_132354.1 | 2 | 470 | ||||
NR_132356.1 | 3 | 318 | ||||
NR_132357.1 | 3 | 318 | ||||
GenBank mRNA | AF080470.1 | - | 3 | 520 | ||
AK057545.1 | - | 3 | 356 | |||
AK128626.1 | - | 4 | 593 | |||
AK315581.1 | - | 3 | 334 | |||
AL596952.1 | - | 3 | 389 | |||
BC004819.1 | - | 3 | 337 | 3450397 | ||
DA798595.1 | - | 2 | 242 | |||
DA859617.1 | - | 3 | 266 | |||
DB212570.1 | - | 2 | 361 | |||
DB289156.1 | - | 3 | 261 |