Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003373.3 | NP_003364.1 | 15 | 2208 | ||
NM_014000.2 | NP_054706.1 | 15 | 2208 | |||
GenBank mRNA | AK126978.1 | - | 15 | 2463 | ||
AK296509.1 | - | 4 | 479 | |||
AK304823.1 | - | 14 | 2137 | |||
AL832130.1 | - | 8 | 1130 | |||
BC039174.1 | - | 15 | 2195 | 4520338 | ||
BX537994.1 | - | 17 | 4918 | |||
EU794673.1 | - | 15 | 2208 | |||
M33308.1 | - | 15 | 2164 |