Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003373.3 | NP_003364.1 | 19 | 3029 | ||
NM_014000.2 | NP_054706.1 | 20 | 3233 | |||
GenBank mRNA | AK126978.1 | - | 19 | 3284 | ||
AK296509.1 | - | 8 | 1300 | |||
AK300280.1 | - | 5 | 810 | |||
AK304823.1 | - | 20 | 3137 | |||
AL832130.1 | - | 13 | 2155 | |||
BC039174.1 | - | 19 | 3016 | 4520338 | ||
BX537994.1 | - | 21 | 5739 | |||
EU794673.1 | - | 20 | 3233 | |||
M33308.1 | - | 19 | 2985 |